4'-Methoxy-3'-(trifluoromethyl)acetophenone 98+%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(4-Methoxy-3-(trifluoromethyl)phenyl)ethanone
PC2631-1G PC2631-25G PC2631-5G
APOSPC2631-1GEA 21.9 EUR
APOSPC2631-1G APOSPC2631-25G APOSPC2631-5G
4'-Methoxy-3'-(trifluoromethyl)acetophenone 98+%
4'-Methoxy-3'-(trifluoromethyl)acetophenone
Formula: C₁₀H₉F₃O₂
MW: 218.18 g/mol
Melting Pt: 56…61 °C
Storage Temperature: Ambient
MDL Number: MFCD01091010
CAS Number: 149105-10-2