1,4-Difluorobenzene 99%
Supplier: Apollo Scientific
Danger
PC2640-500G PC2640-1KG
APOSPC2640-500GEA 195 EUR
APOSPC2640-500G APOSPC2640-1KG
1,4-Difluorobenzene 99%
1,4-Difluorobenzene
Formula: C₆H₄F₂
MW: 114.09 g/mol
Boiling Pt: 88…89 °C
Melting Pt: –13 °C
Density: 1.149 g/cm³ (25 °C)
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00000344
CAS Number: 540-36-3
EINECS: 208-742-9
UN: 1993
ADR: 3,II