2,6-Difluorobenzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2,6-Difluorobenzene-1-sulphonyl chloride
PC2646-100G PC2646-25G
APOSPC2646-100GEA 168 EUR
APOSPC2646-100G APOSPC2646-25G
2,6-Difluorobenzenesulphonyl chloride 97%
2,6-Difluorobenzenesulphonyl chloride
Formula: C₆H₃ClF₂O₂S
Boiling Pt: 105…110 °C/2mmHg
Density: 1.568 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00085004
CAS Number: 60230-36-6
UN: 3265
ADR: 8,II