3-Fluoro-4-iodopyridine 98%
Supplier: Apollo Scientific
Warning
PC2674-1G PC2674-5G
APOSPC2674-1GEA 21.9 EUR
APOSPC2674-1G APOSPC2674-5G
3-Fluoro-4-iodopyridine 98%
3-Fluoro-4-iodopyridine
Formula: C₅H₃FIN
Melting Pt: 85…89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03092927
CAS Number: 22282-75-3