2,6-Difluorobenzonitrile 99%
Supplier: Apollo Scientific
Warning
PC2680-100G PC2680-500G
APOSPC2680-100GEA 21.9 EUR
APOSPC2680-100G APOSPC2680-500G
2,6-Difluorobenzonitrile 99%
2,6-Difluorobenzonitrile
Reaction with guanidine carbonate gives 2,4-diamino-5-fluoroquinazoline, J. Hetero. Chem., 1988, 25, 1173
Formula: C₇H₃F₂N
MW: 139.1 g/mol
Boiling Pt: 197…198 °C
Melting Pt: 25…32 °C
Density: 1.246 g/cm³ (25 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00001778
CAS Number: 1897-52-5
EINECS: 217-589-7
UN: 3439
ADR: 6.1 III