3,4-Difluorobenzonitrile 98%
Supplier: Apollo Scientific
Warning
PC2683-100G PC2683-25G
APOSPC2683-100GEA 37.7 EUR
APOSPC2683-100G APOSPC2683-25G
3,4-Difluorobenzonitrile 98%
3,4-Difluorobenzonitrile
Formula: C₇H₃F₂N
Boiling Pt: 180 °C
Melting Pt: 48…54 °C
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00011666
CAS Number: 64248-62-0
EINECS: 264-751-8
UN: 3439
ADR: 6.1,III