3,5-Difluorobenzonitrile 98%
Supplier: Apollo Scientific
Warning
PC2684-10G PC2684-250G PC2684-50G
APOSPC2684-10GEA 21.9 EUR
APOSPC2684-10G APOSPC2684-250G APOSPC2684-50G
3,5-Difluorobenzonitrile 98%
3,5-Difluorobenzonitrile
Formula: C₇H₃F₂N
Boiling Pt: 166 °C
Melting Pt: 86…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00010311
CAS Number: 64248-63-1
EINECS: 264-752-3
UN: 3439
ADR: 6.1,III