2,6-Difluorobenzophenone 98%
Supplier: Apollo Scientific
Warning
PC2692-100G PC2692-25G PC2692-5G
APOSPC2692-100GEA 364 EUR
APOSPC2692-100G APOSPC2692-25G APOSPC2692-5G
2,6-Difluorobenzophenone 98%
2,6-Difluorobenzophenone
Formula: C₁₃H₈F₂O
MW: 218.2 g/mol
Boiling Pt: 120 °C/2mmHg
Density: 1.244 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009895
CAS Number: 59189-51-4
EINECS: 261-650-0