3,4'-Difluorobenzophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: (3-Fluorophenyl)(4-fluorophenyl)methanone
PC2695-1G PC2695-5G PC2695-10G
APOSPC2695-1GEA 26.1 EUR
APOSPC2695-1G APOSPC2695-5G APOSPC2695-10G
3,4'-Difluorobenzophenone 98%
3,4'-Difluorobenzophenone
Formula: C₁₃H₈F₂O
MW: 218.2000 g/mol
Melting Pt: 56…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00070811
CAS Number: 345-71-1