2,4-Difluorobenzoyl chloride 97%
Supplier: Apollo Scientific
Danger
PC2723-100G PC2723-250G PC2723-25G
APOSPC2723-100GEA 87 EUR
APOSPC2723-100G APOSPC2723-250G APOSPC2723-25G
2,4-Difluorobenzoyl chloride 97%
2,4-Difluorobenzoyl chloride
Formula: C₇H₃ClF₂O
Boiling Pt: 96…98 °C/15mmHg
Density: 1.437 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000658
CAS Number: 72482-64-5
EINECS: 276-690-4
UN: 3265
ADR: 8,II