2-Fluoro-4-iodobenzonitrile
Supplier: Apollo Scientific
|
Warning
|
PC2726-5G
PC2726-25G
APOSPC2726-5GEA
77
EUR
APOSPC2726-5G
APOSPC2726-25G
2-Fluoro-4-iodobenzonitrile
2-Fluoro-4-iodobenzonitrile
Formula:
C₇H₃FIN Melting Pt: 103…105 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094172 CAS Number: 137553-42-5 UN: 3439 ADR: 6.1,III |