2,3-Difluorobenzyl alcohol 98%
Supplier: Apollo Scientific
Danger
PC2729-1G PC2729-5G PC2729-10G
APOSPC2729-1GEA 21.9 EUR
APOSPC2729-1G APOSPC2729-5G APOSPC2729-10G
2,3-Difluorobenzyl alcohol 98%
2,3-Difluorobenzyl alcohol
Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 197
Melting Pt: 9
Density: 1.282 g/cm³ (25 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00010312
CAS Number: 75853-18-8