2,5-Difluorobenzyl alcohol 98%
Supplier: Apollo Scientific
Warning
Synonyms: (2,5-Difluorophenyl)methanol
PC2731-100G PC2731-250G PC2731-25G
APOSPC2731-100GEA 71.2 EUR
APOSPC2731-100G APOSPC2731-250G APOSPC2731-25G
2,5-Difluorobenzyl alcohol 98%
2,5-Difluorobenzyl alcohol
Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 197
Melting Pt: 9
Density: 1.29 g/cm³ (25 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00010627
CAS Number: 75853-20-2
EINECS: 278-325-4