3,4-Difluorobenzylamine 99%
Supplier: Apollo Scientific
Danger
PC2738-100G PC2738-25G PC2738-5G
APOSPC2738-100GEA 203 EUR
APOSPC2738-100G APOSPC2738-25G APOSPC2738-5G
3,4-Difluorobenzylamine 99%
3,4-Difluorobenzylamine
Formula: C₇H₇F₂N
MW: 143.14 g/mol
Density: 1.21 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00010145
CAS Number: 72235-53-1
EINECS: 276-503-6
UN: 2735
ADR: 8,III