3,3'-Difluorobiphenyl 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3,3'-Difluoro-1,1'-biphenyl
PC2748C-1G PC2748C-5G
APOSPC2748C-1GEA 198 EUR
APOSPC2748C-1G APOSPC2748C-5G
3,3'-Difluorobiphenyl 97%
3,3'-Difluorobiphenyl
Formula: C₁₂H₈F₂
MW: 190.19 g/mol
Boiling Pt: 130 °C/14mmHg
Melting Pt: 44415 °C
Density: 1.21 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039216
CAS Number: 396-64-5
EINECS: 206-905-9
UN: 3151
ADR: 9,II