4,4'-Difluorobiphenyl 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4,4-Difluoro-1,1-biphenyl, PFB No. 15
PC2750-5G PC2750-10G PC2750-1G
APOSPC2750-5GEA 52.2 EUR
APOSPC2750-5G APOSPC2750-10G APOSPC2750-1G
4,4'-Difluorobiphenyl 97%
4,4'-Difluorobiphenyl
Formula: C₁₂H₈F₂
MW: 190.19 g/mol
Boiling Pt: 254…255 °C
Melting Pt: 88…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00000349
CAS Number: 398-23-2
EINECS: 206-910-6
UN: 3152
ADR: 9 II
Merck Index: 13,03173