Bis(4-fluorophenyl)sulphone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Fluorophenyl sulphone, 4,4'-Difluoro diphenyl sulphone, 1-Fluor-4-((4-fluorphenyl)sulphonyl)benzol, 4,4'-sulphonylbis(fluorobenzene)
PC2752-100G PC2752-25G PC2752-500G
APOSPC2752-100GEA 78.3 EUR
APOSPC2752-100G APOSPC2752-25G APOSPC2752-500G
Bis(4-fluorophenyl)sulphone 98%
Bis(4-fluorophenyl) sulphone
Formula: C₁₂H₈F₂O₂S
MW: 254.26 g/mol
Melting Pt: 98…100 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000350
CAS Number: 383-29-9
EINECS: 206-847-4