2-Fluoro-5-(trifluoromethyl)benzenesulphonyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: 2-Fluoro-5-(trifluoromethyl)benzene-1-sulphonyl chloride
PC2759-1G PC2759-250MG PC2759-5G
APOSPC2759-1GEA 116 EUR
APOSPC2759-1G APOSPC2759-250MG APOSPC2759-5G
2-Fluoro-5-(trifluoromethyl)benzenesulphonyl chloride
2-Fluoro-5-(trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₃ClF₄O₂S
MW: 262.61 g/mol
Boiling Pt: 100…102 °C/8mmHg
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD01090999
CAS Number: 1744-43-0
UN: 3265
ADR: 8,II