5-Bromo-2-fluoroaniline 98+%
Supplier: Apollo Scientific
Warning
PC2771-100G PC2771-10G PC2771-25G
APOSPC2771-100GEA 59.6 EUR
APOSPC2771-100G APOSPC2771-10G APOSPC2771-25G
5-Bromo-2-fluoroaniline 98+%
5-Bromo-2-fluoroaniline
Formula: C₆H₅BrFN
MW: 190.01 g/mol
Boiling Pt: 105 °C/12mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD03094181
CAS Number: 2924-09-6
UN: 2811
ADR: 6.1 III