2-Bromo-4-fluoro-6-nitrotoluene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-5-fluoro-2-methyl-3-nitrobenzene
PC2783-5G PC2783-25G PC2783-100G
APOSPC2783-5GEA 24.8 EUR
APOSPC2783-5G APOSPC2783-25G APOSPC2783-100G
2-Bromo-4-fluoro-6-nitrotoluene 97%
2-Bromo-4-fluoro-6-nitrotoluene
Formula: C₇H₅BrFNO₂
MW: 234.0225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094189
CAS Number: 502496-33-5