3-Bromo-5-fluoro-2-methylaniline 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-6-bromo-4-fluorotoluene
PC2787-1G PC2787-25G PC2787-5G
APOSPC2787-1GEA 21.9 EUR
APOSPC2787-1G APOSPC2787-25G APOSPC2787-5G
3-Bromo-5-fluoro-2-methylaniline 98%
3-Bromo-5-fluoro-2-methylaniline
Formula: C₇H₇BrFN
MW: 204.0400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094193
CAS Number: 502496-36-8