2-Fluorothioanisole
Supplier: Apollo Scientific
Warning
Synonyms: 1-Fluoro-2-(methylthio)benzene, 2-Fluorophenyl methyl sulphide
PC2793-25G PC2793-5G
APOSPC2793-25GEA 89.9 EUR
APOSPC2793-25G APOSPC2793-5G
2-Fluorothioanisole
2-Fluorothioanisole
Formula: C₇H₇FS
MW: 142.2 g/mol
Boiling Pt: 197…199 °C
Density: 1.174 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD03093762
CAS Number: 655-20-9