2-Fluoro-3-(4-methoxyphenyl)-3-oxopropanenitrile, Technical Grade
Supplier: Apollo Scientific
PC28217-1G PC28217-5G
APOSPC28217-1GEA 180 EUR
APOSPC28217-1G APOSPC28217-5G
2-Fluoro-3-(4-methoxyphenyl)-3-oxopropanenitrile, Technical Grade
2-Fluoro-3-(4-methoxyphenyl)-3-oxopropanenitrile
Formula: C₁₀H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD18252980
CAS Number: 1280787-24-7