5-Fluoro-4-hydroxypyrimidine
Supplier: Apollo Scientific
Warning
PC2823-1G PC2823-25G PC2823-5G PC2823-100G
APOSPC2823-1GEA 14.7 EUR
APOSPC2823-1G APOSPC2823-25G APOSPC2823-5G APOSPC2823-100G
5-Fluoro-4-hydroxypyrimidine
5-Fluoro-4-hydroxypyrimidine
Formula: C4H3FN2O
MW: 114.778 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00223674
CAS Number: 671-35-2