3',5'-Difluoro-4'-hydroxypropiophenone 98%
Supplier: Apollo Scientific
Warning
PC2828-1G PC2828-5G PC2828-25G
APOSPC2828-1GEA 42.2 EUR
APOSPC2828-1G APOSPC2828-5G APOSPC2828-25G
3',5'-Difluoro-4'-hydroxypropiophenone 98%
Formula: C₉H₈F₂O₂
Melting Pt: 128…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00061287
CAS Number: 178374-78-2