4-(Difluoromethoxy)aniline 96%
Supplier: Apollo Scientific
Warning
PC2846E-5G PC2846E-100G PC2846E-25G
APOSPC2846E-5GEA 21.9 EUR
APOSPC2846E-5G APOSPC2846E-100G APOSPC2846E-25G
4-(Difluoromethoxy)aniline 96%
4-(Difluoromethoxy)aniline
Formula: C₇H₇F₂NO
MW: 159.14 g/mol
Boiling Pt: 231 °C
Density: 1.286 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00085005
CAS Number: 22236-10-8
UN: 2810
ADR: 6.1,III