4-(Difluoromethoxy)benzaldehyde 98%
Supplier: Apollo Scientific
Warning
PC2847D-25G PC2847D-5G PC2847D-100G
APOSPC2847D-25GEA 72.5 EUR
APOSPC2847D-25G APOSPC2847D-5G APOSPC2847D-100G
4-(Difluoromethoxy)benzaldehyde 98%
4-(Difluoromethoxy)benzaldehyde
Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
Boiling Pt: 58…60 °C/0.5mmHg
Density: 1.295 g/cm³ (25 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00042252
CAS Number: 73960-07-3