4-(Difluoromethoxy)benzoic acid 98%
Supplier: Apollo Scientific
Warning
PC2847K-5G PC2847K-1G
APOSPC2847K-5GEA 37.7 EUR
APOSPC2847K-5G APOSPC2847K-1G
4-(Difluoromethoxy)benzoic acid 98%
4-(Difluoromethoxy)benzoic acid
Formula: C₈H₆F₂O₃
MW: 188.13 g/mol
Melting Pt: 165…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00221454
CAS Number: 4837-20-1