α-Bromo-4-(difluoromethoxy)toluene 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1-(Bromomethyl)-4-(difluoromethoxy)benzene, p-(Difluoromethoxy)benzyl bromide, 4-(Difluoromethoxy)benzyl bromide
PC2847Q-1G PC2847Q-25G PC2847Q-5G
APOSPC2847Q-1GEA 30.6 EUR
APOSPC2847Q-1G APOSPC2847Q-25G APOSPC2847Q-5G
α-Bromo-4-(difluoromethoxy)toluene 98%
α-Bromo-4-(difluoromethoxy)toluene
Formula: C₈H₇BrF₂O
MW: 237.04 g/mol
Boiling Pt: 64…66 °C/0.3mmHg
Storage Temperature: Ambient
MDL Number: MFCD00085007
CAS Number: 3447-53-8
UN: 3265
ADR: 8 III