1-Bromo-2-(difluoromethoxy)benzene 97%
Supplier: Apollo Scientific
Warning
PC2847QM-100G PC2847QM-25G PC2847QM-5G
APOSPC2847QM-100GEA 173 EUR
APOSPC2847QM-100G APOSPC2847QM-25G APOSPC2847QM-5G
1-Bromo-2-(difluoromethoxy)benzene 97%
1-Bromo-2-(difluoromethoxy)benzene
Formula: C₇H₅BrF₂O
MW: 223.02 g/mol
Boiling Pt: 85…87 °C/15mmHg
Density: 1.6 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00221468
CAS Number: 175278-33-8