1-(Difluoromethoxy)-2-nitrobenzene 97%
Supplier: Apollo Scientific
Warning
PC2847RS-100G PC2847RS-25G PC2847RS-5G
APOSPC2847RS-100GEA 404 EUR
APOSPC2847RS-100G APOSPC2847RS-25G APOSPC2847RS-5G
1-(Difluoromethoxy)-2-nitrobenzene 97%
1-(Difluoromethoxy)-2-nitrobenzene
Formula: C₇H₅F₂NO₃
MW: 189.12 g/mol
Boiling Pt: 73…75 °C/0.3mmHg
Storage Temperature: Ambient
MDL Number: MFCD00221470
CAS Number: 22225-77-0