1-(Difluoromethoxy)-4-nitrobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Difluoromethoxy)nitrobenzene
PC2847S-100G PC2847S-25G PC2847S-5G
APOSPC2847S-100GEA 195 EUR
APOSPC2847S-100G APOSPC2847S-25G APOSPC2847S-5G
1-(Difluoromethoxy)-4-nitrobenzene 98%
1-(Difluoromethoxy)-4-nitrobenzene
Formula: C₇H₅F₂NO₃
MW: 189.12 g/mol
Boiling Pt: 108…110 °C/1.5mmHg
Melting Pt: 33…39 °C
Storage Temperature: Ambient
MDL Number: MFCD00085009
CAS Number: 1544-86-1