2-Bromo-4'-(difluoromethoxy)acetophenone 97%
Supplier: Apollo Scientific
Danger
Synonyms: 4-(Difluoromethoxy)phenacyl bromide
PC2847T-1G PC2847T-25G PC2847T-5G
APOSPC2847T-1GEA 21.9 EUR
APOSPC2847T-1G APOSPC2847T-25G APOSPC2847T-5G
2-Bromo-4'-(difluoromethoxy)acetophenone 97%
2-Bromo-4'-(difluoromethoxy)acetophenone
Formula: C₉H₇BrF₂O₂
Melting Pt: 63…68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052200
CAS Number: 141134-24-9
UN: 3261
ADR: 8 III