1,3-Difluoro-4-nitrobenzene 99%
Supplier: Apollo Scientific
Danger
Synonyms: 2,4-Difluoro-1-nitrobenzene, 2,4-Difluoronitrobenzene
PC2850-500G PC2850-100G PC2850-1KG
APOSPC2850-500GEA 40.6 EUR
APOSPC2850-500G APOSPC2850-100G APOSPC2850-1KG
1,3-Difluoro-4-nitrobenzene 99%
1,3-Difluoro-4-nitrobenzene
Formula: C₆H₃F₂NO₂
Boiling Pt: 203…204 °C
Melting Pt: 10 °C
Density: 1.451 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00007050
CAS Number: 446-35-5
EINECS: 207-167-0
UN: 2810
ADR: 6.1,III