1,2-Difluoro-4-nitrobenzene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 3,4-Difluoronitrobenzene
PC2862-100G PC2862-500G
APOSPC2862-100GEA 21.9 EUR
APOSPC2862-100G APOSPC2862-500G
1,2-Difluoro-4-nitrobenzene 99%
1,2-Difluoro-4-nitrobenzene
Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 76…80 °C/11mmHg
Density: 1.437 g/cm³ (25 °C)
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00007198
CAS Number: 369-34-6
EINECS: 206-718-2