2,6-Difluorophenol 98%
Supplier: Apollo Scientific
Danger
PC2870-100G PC2870-25G PC2870-5G
APOSPC2870-100GEA 224 EUR
APOSPC2870-100G APOSPC2870-25G APOSPC2870-5G
2,6-Difluorophenol 98%
2,6-Difluorophenol
Formula: C₆H₄F₂O
MW: 130.09 g/mol
Boiling Pt: 59…61 °C/17mmHg
Melting Pt: 38…41 °C
Density: 1.27 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00002158
CAS Number: 28177-48-2
EINECS: 248-884-9
UN: 2925
ADR: 4.1+8,III