3,4-Difluorophenol 99%
Supplier: Apollo Scientific
Danger
PC2871-100G PC2871-25G PC2871-5G
APOSPC2871-100GEA 127 EUR
APOSPC2871-100G APOSPC2871-25G APOSPC2871-5G
3,4-Difluorophenol 99%
3,4-Difluorophenol
Formula: C₆H₄F₂O
MW: 130.09 g/mol
Boiling Pt: 84…86 °C/20mmHg
Melting Pt: 34…38 °C
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00010315
CAS Number: 2713-33-9
EINECS: 424-300-1
UN: 2925
ADR: 4.1+8,III