(3,4-Difluorophenyl)acetic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3,4-Difluorophenylacetic acid
PC2874E-100G PC2874E-25G PC2874E-5G
APOSPC2874E-100GEA 143 EUR
APOSPC2874E-100G APOSPC2874E-25G APOSPC2874E-5G
(3,4-Difluorophenyl)acetic acid 98%
(3,4-Difluorophenyl)acetic acid
Formula: (C₆H₃F₂)CH₂COOH
Melting Pt: 47…51 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00010002
CAS Number: 658-93-5
EINECS: 211-527-2