1-(2,4-Difluorophenyl)biguanide hydrochloride 97%
Supplier: Apollo Scientific
Warning
PC2875-10G PC2875-1G PC2875-5G
APOSPC2875-10GEA 208 EUR
APOSPC2875-10G APOSPC2875-1G APOSPC2875-5G
1-(2,4-Difluorophenyl)biguanide hydrochloride 97%
1-(2,4-Difluorophenyl)biguanide hydrochloride
Formula: C₈H₁₀ClF₂N₅
Melting Pt: 226…228 °C
Storage Temperature: Ambient
MDL Number: MFCD00053015
CAS Number: 66088-52-6