1,3-Difluoro-2-propanol 97%
Supplier: Apollo Scientific
Warning
PC2878-25G PC2878-5G
APOSPC2878-25GEA 255 EUR
APOSPC2878-25G APOSPC2878-5G
1,3-Difluoro-2-propanol 97%
1,3-Difluoro-2-propanol
Formula: C₃H₆F₂O
MW: 96.08 g/mol
Boiling Pt: 127 °C
Density: 1.24 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00000452
CAS Number: 453-13-4
EINECS: 207-216-6
UN: 1987
ADR: 3,III