1-Bromo-5-fluoro-2-nitrobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-4-fluoro-1-nitrobenzene, 2-Bromo-4-fluoronitrobenzene
PC2911-5G PC2911-25G PC2911-100G
APOSPC2911-5GEA 21.9 EUR
APOSPC2911-5G APOSPC2911-25G APOSPC2911-100G
1-Bromo-5-fluoro-2-nitrobenzene 98%
1-Bromo-5-fluoro-2-nitrobenzene
Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Boiling Pt: 70…75 °C/0.4mmHg
Melting Pt: 39…42 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00792441
CAS Number: 700-36-7