5-Bromo-2-fluorophenol 98%
Supplier: Apollo Scientific
Warning
PC2914-25G PC2914-5G PC2914-100G PC2914-500G
APOSPC2914-25GEA 43.5 EUR
APOSPC2914-25G APOSPC2914-5G APOSPC2914-100G APOSPC2914-500G
5-Bromo-2-fluorophenol 98%
5-Bromo-2-fluorophenol
Formula: C₆H₄BrFO
MW: 191 g/mol
Boiling Pt: 53 °C/2mmHg
Storage Temperature: Ambient
MDL Number: MFCD07782066
CAS Number: 112204-58-7