2-Fluoro-4-iodopyridine
Supplier: Apollo Scientific
Warning
PC2918-100G PC2918-25G
APOSPC2918-100GEA 394 EUR
APOSPC2918-100G APOSPC2918-25G
2-Fluoro-4-iodopyridine
2-Fluoro-4-iodopyridine
Formula: C₅H₃FIN
MW: 222.99 g/mol
Melting Pt: 57…59 °C
Storage Temperature: Ambient
MDL Number: MFCD03092926
CAS Number: 22282-70-8