5-Fluoro-3-piperidin-4-yl-1,2-benzisoxazole hydrochloride 96%
Supplier: Apollo Scientific
Warning
PC2919-1G PC2919-5G
APOSPC2919-1GEA 143 EUR
APOSPC2919-1G APOSPC2919-5G
5-Fluoro-3-piperidin-4-yl-1,2-benzisoxazole hydrochloride 96%
5-Fluoro-3-piperidin-4-yl-1,2-benzisoxazole hydrochloride
Formula: C₁₂H₁₄ClFN₂O
MW: 256.71 g/mol
Melting Pt: 300…301 °C
Storage Temperature: Ambient
MDL Number: MFCD06200879
CAS Number: 84163-16-6