2,5-Difluorotoluene 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1,4-Difluoro-2-methylbenzene
PC2940-10G PC2940-25G
APOSPC2940-10GEA 21.9 EUR
APOSPC2940-10G APOSPC2940-25G
2,5-Difluorotoluene 98%
2,5-Difluorotoluene
Formula: C₇H₆F₂
MW: 128.12 g/mol
Boiling Pt: 115…117 °C
Melting Pt: –35 °C
Density: 1.36 g/cm³ (25 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00000347
CAS Number: 452-67-5
UN: 2388
ADR: 3,II