2,6-Difluorotoluene
Supplier: Apollo Scientific
Danger
Synonyms: 1,3-Difluoro-2-methylbenzene
PC2941-1G PC2941-25G PC2941-5G
APOSPC2941-1GEA 56.8 EUR
APOSPC2941-1G APOSPC2941-25G APOSPC2941-5G
2,6-Difluorotoluene
2,6-Difluorotoluene
Formula: C₇H₆F₂
MW: 128.12 g/mol
Boiling Pt: 111…112 °C
Density: 1.129 g/cm³ (25 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00043898
CAS Number: 443-84-5
UN: 2388
ADR: 3,II