3,4-Difluorotoluene 99%
Supplier: Apollo Scientific
Warning
PC2942-100G PC2942-25G PC2942-5G
APOSPC2942-100GEA 335 EUR
APOSPC2942-100G APOSPC2942-25G APOSPC2942-5G
3,4-Difluorotoluene 99%
3,4-Difluorotoluene
Formula: C₇H₆F₂
MW: 128.12 g/mol
Boiling Pt: 110…113 °C
Density: 1.12 g/cm³ (25 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00075087
CAS Number: 2927-34-6
UN: 1993
ADR: 3,III