2,5-Bis(trifluoromethyl)benzenesulphonyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: 2,5-Bis(trifluoromethyl)benzene-1-sulphonyl chloride
PC2948-25G PC2948-5G
APOSPC2948-25GEA 102 EUR
APOSPC2948-25G APOSPC2948-5G
2,5-Bis(trifluoromethyl)benzenesulphonyl chloride
2,5-Bis(trifluoromethyl)benzenesulphonyl chloride
Formula: C₈H₃ClF₆O₂S
Melting Pt: 59…63 °C
Storage Temperature: Ambient
MDL Number: MFCD03094042
CAS Number: 351003-22-0
UN: 3261
ADR: 8,II