3-Fluoro-4-nitroanisole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-4-methoxy-1-nitrobenzene
PC2959-25G PC2959-5G PC2959-100G
APOSPC2959-25GEA 55.1 EUR
APOSPC2959-25G APOSPC2959-5G APOSPC2959-100G
3-Fluoro-4-nitroanisole 98%
3-Fluoro-4-nitroanisole
Formula: C₇H₆FNO₃
MW: 171.13 g/mol
Melting Pt: 56…58 °C
Storage Temperature: Ambient
MDL Number: MFCD04115632
CAS Number: 446-38-8