3-Nitro-5-(trifluoromethyl)phenol
Supplier: Apollo Scientific
Warning
PC2962-1G PC2962-5G
APOSPC2962-1GEA 21.9 EUR
APOSPC2962-1G APOSPC2962-5G
3-Nitro-5-(trifluoromethyl)phenol
3-Nitro-5-(trifluoromethyl)phenol
Formula: C₇H₄F₃NO₃
Melting Pt: 89…92 °C
Storage Temperature: Ambient
MDL Number: MFCD04973779
CAS Number: 349-57-5